Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098799.1 | NP_001092269.1 | 1, 2 | 139 | ||
GenBank mRNA | BB649411.1 | - | 1, 2 | 139 |