Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033373.2 | NP_001028545.2 | 4 | 444 | ||
GenBank mRNA | BC147314.1 | - | 4 | 444 | 9056364 | |
BC147315.1 | - | 4 | 444 | 9056370 | ||
BC147758.1 | - | 4 | 444 | 9087483 | ||
BC147763.1 | - | 4 | 444 | 9087488 | ||
BC172036.1 | - | 4 | 441 | 9054322 |