Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080550.2 | NP_001074019.1 | 8 | 1758 | ||
NM_001276432.1 | NP_001263361.1 | 7 | 2444 | |||
GenBank mRNA | AK136599.1 | - | 7 | 2449 | ||
AY757956.1 | - | 8 | 1758 |