Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080285.1 | NP_525024.1 | 16 | 3922 | ||
GenBank mRNA | AK032356.1 | - | 14 | 2303 | ||
AK081137.1 | - | 12 | 2386 | |||
AK173254.1 | - | 16 | 3828 | |||
BC068156.1 | - | 16 | 3683 | |||
BC141407.1 | - | 16 | 3918 | 9056020 |