Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017316.2 | NP_059012.2 | 16 | 1947 | ||
XM_006235067.2 | XP_006235129.1 | 16 | 2744 | |||
XM_006235068.2 | XP_006235130.1 | 17 | 2747 | |||
XM_006235069.2 | XP_006235131.1 | 14 | 4189 | |||
GenBank mRNA | AF080453.1 | - | 16 | 1947 |