Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50352
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
488
6921326
FQ211494.1
-
488
FQ211970.1
-
484
FQ212357.1
-
485
FQ212579.1
-
484
FQ212640.1
-
484
FQ212789.1
-
488
FQ212869.1
-
488
FQ213028.1
-
484
FQ213029.1
-
482
FQ213091.1
-
484
FQ213115.1
-
490
FQ213200.1
-
487
FQ213216.1
-
484
FQ213310.1
-
377
FQ213366.1
-
483
FQ213496.1
-
487
FQ213520.1
-
488
FQ213526.1
-
484
FQ213581.1
-
484
FQ213598.1
-
484
FQ213637.1
-
484
FQ213695.1
-
484
FQ213703.1
-
484
FQ213760.1
-
484
FQ213762.1
-
489
FQ213913.1
-
484
FQ213941.1
-
488
FQ213981.1
-
485
FQ213985.1
-
378
FQ213999.1
-
483
FQ214059.1
-
488
FQ214181.1
-
488
FQ214262.1
-
488
FQ214491.1
-
484
FQ214500.1
-
484
FQ214519.1
-
484
FQ214525.1
-
484
FQ214540.1
-
488
FQ215317.1
-
484
FQ215600.1
-
488
FQ216397.1
-
484
FQ216471.1
-
485
FQ218091.1
-
484
FQ220250.1
-
484
FQ220325.1
-
484
FQ220698.1
-
488
FQ220779.1
-
484
FQ222184.1
-
487
FQ223357.1
-
488
FQ225272.1
-
484
FQ226994.1
-
488
FQ229228.1
-
484
FQ229580.1
-
488
FQ229619.1
-
484
FQ229790.1
-
484
FQ229905.1
-
483
FQ231022.1
-
488
FQ231702.1
-
488
FQ233337.1
-
487
FQ234377.1
-
488
FQ235266.1
-
485
M17069.1
-
486
M19312.1
-
467
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007589, BC100301, AK013068, BC021347, AK012247, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0