Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037713.4 | NP_001032802.2 | 5 | 533 | ||
NM_001291153.1 | NP_001278082.1 | 3 | 248 | |||
GenBank mRNA | AK046736.1 | - | 4 | 304 | ||
AK140315.1 | - | 4 | 2456 | |||
BY726623.1 | - | 3 | 248 | |||
CK130151.1 | - | 4 | 468 | |||
EL957551.1 | - | 3 | 238 |