Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045527.1 | NP_001038992.1 | 6 | 2172 | ||
GenBank mRNA | AK136195.1 | - | 3 | 638 | ||
AK166561.1 | - | 3 | 614 |