Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033539.2 | NP_001028711.1 | 3 | 535 | ||
GenBank mRNA | AK139543.1 | - | 2 | 466 | ||
AK157452.1 | - | 3 | 536 | |||
AK163328.1 | - | 2 | 418 | |||
AY833559.1 | - | 3 | 535 | |||
BC132130.1 | - | 1 | 305 | 40130407 | ||
BC132132.1 | - | 1 | 305 | 40130409 |