Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081190.1 | NP_001074659.1 | 9 | 1254 | ||
GenBank mRNA | AK141642.1 | - | 5 | 679 | ||
BC151105.1 | - | 9 | 1304 | 9088018 |