Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033409.3 | NP_001028581.1 | 18 | 3508 | ||
GenBank mRNA | AK040883.1 | - | 1 | 1110 | ||
AK052873.1 | - | 13 | 2709 | |||
AK085901.1 | - | 18 | 3470 | |||
AK137380.1 | - | 18 | 3509 | |||
BC026896.1 | - | 3 | 1850 |