Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029264.2 | NP_083540.1 | 4 | 413 | ||
GenBank mRNA | AK014686.1 | - | 3 | 791 | ||
AK133079.1 | - | 4 | 414 | |||
AM690754.1 | - | 1 | 297 |