Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dock1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033420.2 | NP_001028592.1 | 9 | 931 | ||
GenBank mRNA | AK082835.1 | - | 9 | 927 | ||
AK138143.1 | - | 9 | 878 | |||
AK141224.1 | - | 9 | 927 | |||
AK147836.1 | - | 9 | 929 | |||
AK163853.1 | - | 9 | 932 | |||
AK164963.1 | - | 9 | 913 |