Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081354.2 | NP_001074823.2 | 7 | 2719 | ||
NM_001256048.1 | NP_001242977.1 | 6 | 2608 |