Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045529.3 | NP_001038994.2 | 7 | 1048 | ||
GenBank mRNA | AK132226.1 | - | 7 | 904 | ||
AK144885.1 | - | 8 | 1003 | |||
AK172896.1 | - | 7 | 1048 | |||
BC098072.1 | - | 7 | 889 | 30109159 | ||
BC145705.1 | - | 7 | 997 | 40131008 |