Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019141.2 | NP_062014.2 | 2, 3 | 311 | ||
XM_006231128.2 | XP_006231190.1 | 2, 3 | 922 | |||
XM_006231129.2 | XP_006231191.1 | 2, 3 | 922 | |||
XM_008760286.1 | XP_008758508.1 | 2, 3 | 922 | |||
GenBank mRNA | CB717588.1 | - | 1, 2 | 36 | ||
L14463.1 | - | 2, 3 | 307 |