Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139221.1 | - | 6 | 1408 | ||
AK164883.1 | - | 5 | 2750 | |||
BC064128.1 | - | 7 | 1577 |