Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019185.1 | NP_062058.1 | 4 | 1130 | ||
GenBank mRNA | L22760.1 | - | 4 | 1130 |