Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080924.2 | NP_001074393.1 | 9 | 5363 | ||
NM_001290501.1 | NP_001277430.1 | 9 | 6256 | |||
GenBank mRNA | AK052287.1 | - | 1 | 2334 | ||
AK142465.1 | - | 1 | 300 |