Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024626.2 | NP_001019797.2 | 3 | 2126 | ||
GenBank mRNA | AK143655.1 | - | 4 | 1627 | ||
BC062115.1 | - | 1 | 2153 | |||
BC090848.1 | - | 4 | 1774 | 30626866 | ||
BC096521.1 | - | 2 | 1918 |
Gene Symbol: A130033P14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037654.1 | - | 1 | 2457 |