Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyg11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033634.3 | NP_001028806.2 | 14 | 5178 | ||
GenBank mRNA | AK013433.1 | - | 1 | 436 | ||
AK122545.1 | - | 7 | 3567 | |||
BC020312.2 | - | 1 | 2336 | |||
BC117977.2 | - | 8 | 3031 | 40099428 | ||
BC117978.2 | - | 8 | 3031 | 40099429 |