Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033488.2 | NP_001028660.2 | 14 | 1713 | ||
GenBank mRNA | AK133061.1 | - | 11 | 2615 | ||
AK156138.1 | - | 15 | 1866 | |||
AK157892.1 | - | 12 | 2396 | |||
AK162620.1 | - | 12 | 1458 | |||
AK171660.1 | - | 9 | 2077 | |||
AK172423.1 | - | 14 | 1714 |