Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034882.3 | NP_001030054.3 | 1 | 2764 | ||
GenBank mRNA | AK159684.1 | - | 1 | 2765 | ||
AK163295.1 | - | 2 | 2722 |