Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar7f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010839.1 | NP_001010839.1 | 11 | |||
GenBank mRNA | BC139175.1 | - | 1 | 39 | 8862197 | |
BC139176.1 | - | 1 | 39 | 8862198 |
Set Membership: |
G-protein coupled receptor |