Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dupd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013826.2 | NP_001013848.1 | 4 | 1085 | ||
GenBank mRNA | AK075665.1 | - | 4 | 1086 |