Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081069.1 | NP_001074538.1 | 17 | 1804 | ||
GenBank mRNA | AF061934.1 | - | 16 | 1709 | ||
AK019054.1 | - | 1 | 161 | |||
AK039314.1 | - | 16 | 2426 | |||
AK080707.1 | - | 17 | 1898 | |||
AK161551.1 | - | 13 | 1863 | |||
AK166716.1 | - | 3 | 1049 | |||
BC019741.1 | - | 6 | 1136 | |||
BC049910.1 | - | 15 | 2508 |