Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnlz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139503.1 | NP_001132975.1 | 2 | 2435 | ||
NM_001139504.1 | NP_001132976.1 | 3 | 2579 | |||
NM_026828.3 | NP_081104.1 | 3 | 2575 | |||
GenBank mRNA | AK142070.1 | - | 3 | 2576 | ||
BC031595.1 | - | 2 | 2342 |