Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034905.2 | NP_001030077.1 | 2 | 1715 | ||
NM_001039563.3 | NP_001034652.2 | 3 | 617 | |||
NM_001286493.1 | NP_001273422.1 | 3 | 620 | |||
GenBank mRNA | AK136627.1 | - | 2 | 1716 |