Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039173.1 | NP_001034262.1 | 1, 2 | 69 | ||
GenBank mRNA | AB221044.1 | - | 1, 2 | 69 | ||
BC138955.1 | - | 1, 2 | 120 | 8861977 | ||
BC138957.1 | - | 1, 2 | 120 | 8861979 |