Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025554.2 | NP_079830.2 | 7 | 993 | ||
GenBank mRNA | AK010567.1 | - | 7 | 1001 | ||
AK015823.1 | - | 7 | 991 | |||
AK088610.1 | - | 7 | 1050 | |||
AK132109.1 | - | 7 | 1048 | |||
AK144668.1 | - | 7 | 1054 | |||
AK145201.1 | - | 7 | 1059 | |||
BC005599.1 | - | 7 | 1045 | |||
BC026842.1 | - | 5 | 1221 | 4210827 | ||
BC037681.1 | - | 4 | 2388 | |||
BC045521.1 | - | 7 | 993 | 2647754 |