Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019187.1 | NP_062060.1 | 4 | 456 | ||
GenBank mRNA | BC081811.1 | - | 621 | |||
BC098666.1 | - | 614 | ||||
BC126094.1 | - | 628 | 8370738 | |||
FQ216025.1 | - | 668 | ||||
FQ230994.1 | - | 2 | 233 | |||
L20427.1 | - | 4 | 456 |