Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025691.1 | NP_079967.1 | 16 | 1630 | ||
GenBank mRNA | AK017869.1 | - | 2 | 186 | ||
AK133784.1 | - | 16 | 1631 | |||
BC003434.1 | - | 16 | 1618 | |||
BC019196.1 | - | 16 | 1623 | |||
BC089462.1 | - | 7 | 638 |