Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm8909 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081032.2 | NP_001074501.2 | 8 | 1319 | ||
GenBank mRNA | AB359231.2 | - | 8 | 1319 |