Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025776.2 | NP_080052.1 | 11 | 2025 | ||
GenBank mRNA | AK018452.1 | - | 11 | 2048 | ||
AK045116.1 | - | 11 | 2046 | |||
AK150271.1 | - | 11 | 2038 | |||
AK151758.1 | - | 11 | 2038 | |||
AK152252.1 | - | 11 | 2038 | |||
AK166600.1 | - | 11 | 2046 | |||
AK167509.1 | - | 11 | 2065 | |||
AK168919.1 | - | 11 | 2026 | |||
BC042691.1 | - | 12 | 1961 | |||
BC080205.1 | - | 11 | 2023 | 3709760 |