Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100187.1 | NP_001093657.1 | 2, 3 | 238 | ||
GenBank mRNA | AK132492.1 | - | 2, 3 | 238 | ||
BC151030.1 | - | 223 | 9087943 |