Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033136.3 | NP_001028308.1 | 13 | 2211 | ||
GenBank mRNA | AK004786.1 | - | 13 | 2137 | ||
AK151090.1 | - | 13 | 2132 | |||
AK153322.1 | - | 13 | 2132 | |||
AK153369.1 | - | 13 | 2131 | |||
AK171770.1 | - | 13 | 2211 | |||
BC055754.1 | - | 13 | 2105 |