Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026605.2 | NP_080881.2 | 3 | 309 | ||
GenBank mRNA | AK146894.1 | - | 3 | 291 | ||
AK155093.1 | - | 3 | 310 | |||
AK155375.1 | - | 3 | 264 | |||
AK165591.1 | - | 3 | 288 | |||
AK166290.1 | - | 264 | ||||
AK168541.1 | - | 3 | 291 | |||
AK168757.1 | - | 3 | 291 | |||
AK172040.1 | - | 3 | 264 | |||
BC049852.1 | - | 3 | 254 | 4163760 |