Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dydc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027094.1 | NP_081370.1 | 4 | 427 | ||
GenBank mRNA | AK006511.1 | - | 4 | 427 | ||
BC049632.1 | - | 4 | 378 | 6771945 |