Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033150.1 | NP_001028322.1 | 4, 5 | 565 | ||
GenBank mRNA | AK122386.1 | - | 649 | |||
AK156652.1 | - | 1, 2 | 187 | |||
AK185623.1 | - | 2, 3 | 155 | |||
AK219223.1 | - | 3, 4 | 230 | |||
BC058984.1 | - | 2, 3 | 156 | |||
CX240883.1 | - | 4 | 565 |