Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027410.1 | NP_081686.1 | 4 | 581 | ||
GenBank mRNA | AB093295.1 | - | 3 | 750 | ||
AK032858.1 | - | 4 | 556 | |||
AK034020.1 | - | 4 | 556 | |||
AK163522.1 | - | 3 | 810 | |||
AK169888.1 | - | 4 | 528 | |||
AK170108.1 | - | 4 | 528 | |||
BC046968.1 | - | 4 | 581 | 6306206 |