Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgrmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027558.1 | NP_081834.1 | 3 | 1670 | ||
GenBank mRNA | AK014543.1 | - | 3 | 1569 | ||
AK140064.1 | - | 3 | 1650 | |||
AK165560.1 | - | 1 | 853 | |||
AK184626.1 | - | 1 | 241 | |||
AX775753.1 | - | 3 | 1626 | |||
BC044759.2 | - | 3 | 1611 |