Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spdya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142631.1 | NP_001136103.1 | 2 | 149 | ||
NM_029254.1 | NP_083530.1 | 2 | 149 | |||
GenBank mRNA | AK014910.1 | - | 2 | 139 | ||
AK077138.1 | - | 2 | 150 | |||
AY820306.1 | - | 1 | 50 | |||
AY820307.1 | - | 1 | 50 | |||
BC147706.1 | - | 2 | 127 | 9087465 | ||
BC147719.1 | - | 2 | 127 | 9087478 |
Gene Symbol: Mettl4-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016064.1 | - | 1088 |