Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027643.1 | NP_081919.1 | 5 | 580 | ||
GenBank mRNA | AK016504.1 | - | 5 | 541 | ||
AK033510.1 | - | 5 | 402 | |||
AK033896.1 | - | 5 | 411 | |||
AK035418.1 | - | 4 | 1298 | |||
AK162625.1 | - | 5 | 414 | |||
AK170548.1 | - | 5 | 406 | |||
AK187772.1 | - | 1 | 235 | |||
AK193575.1 | - | 2 | 155 | |||
AK209302.1 | - | 1 | 237 | |||
AK213481.1 | - | 4 | 307 | |||
AK216197.1 | - | 4 | 308 | |||
BC020020.1 | - | 5 | 431 | 3156924 | ||
BC058212.1 | - | 1 | 97 |