Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027908.1 | NP_082184.1 | 5 | 776 | ||
GenBank mRNA | AK005066.1 | - | 5 | 732 | ||
AK090143.1 | - | 5 | 767 | |||
BC022626.1 | - | 5 | 654 | |||
BC043681.1 | - | 5 | 709 | |||
BC055025.1 | - | 5 | 674 | |||
BC118616.1 | - | 5 | 763 | 40107296 | ||
BC121826.1 | - | 5 | 762 | 40107295 |