Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028128.1 | NP_082404.1 | 6 | 499 | ||
GenBank mRNA | AK011489.1 | - | 6 | 510 | ||
AK150036.1 | - | 512 | ||||
AK170211.1 | - | 6 | 517 | |||
AK171372.1 | - | 499 | ||||
BC023674.1 | - | 5 | 437 | |||
BC089001.1 | - | 6 | 499 | 6415420 |