Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028462.1 | NP_082738.1 | 1 | 966 | ||
GenBank mRNA | AK014242.1 | - | 1 | 844 | ||
BC049157.1 | - | 1 | 1065 | |||
BC119565.1 | - | 1 | 966 | 40057718 | ||
BC119566.1 | - | 1 | 966 | 40057721 |