Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028479.1 | NP_082755.1 | 3, 4 | 704 | ||
GenBank mRNA | AK005551.1 | - | 2, 3 | 344 | ||
AK052575.1 | - | 2, 3 | 359 | |||
AK217601.1 | - | 1, 2 | 121 | |||
BC054431.1 | - | 2, 3 | 291 | |||
BC110993.1 | - | 2, 3 | 310 | 5363496 | ||
BC116732.1 | - | 3, 4 | 704 | 40126051 | ||
BC116734.1 | - | 3, 4 | 704 | 40126053 |