Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081164.1 | NP_001074633.1 | 21 | 3897 | ||
NM_001256033.1 | NP_001242962.1 | 21 | 3900 | |||
GenBank mRNA | AK034477.1 | - | 1 | 273 | ||
AK078157.1 | - | 1 | 93 | |||
AK122429.1 | - | 16 | 3191 | |||
BC042641.1 | - | 1 | 638 | |||
BC138373.1 | - | 21 | 3872 | 8861393 | ||
BC145259.1 | - | 21 | 3869 | 9053812 |