Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103156.2 | NP_001096626.1 | 6 | 7248 | ||
NM_001103157.2 | NP_001096627.1 | 6 | 7603 | |||
NM_001285469.1 | NP_001272398.1 | 6 | 7157 | |||
NM_001285470.1 | NP_001272399.1 | 5 | 7122 | |||
NM_001285471.1 | NP_001272400.1 | 5 | 1531 | |||
NM_028734.5 | NP_083010.2 | 6 | 7305 | |||
GenBank mRNA | AK089927.1 | - | 5 | 1505 |