Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028770.2 | NP_083046.2 | 4 | 675 | ||
GenBank mRNA | AK004811.2 | - | 3 | 464 | ||
AK036900.1 | - | 4 | 714 | |||
AK171114.1 | - | 564 | ||||
BC119366.1 | - | 4 | 675 | 8734115 | ||
BC120601.1 | - | 4 | 675 | 40129524 |